Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Amino-2-chloro-3-picoline, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
38186-82-2
C6H7ClN2
142.59
VSBISZPNLZFTPG-UHFFFAOYSA-N
5-amino-2-chloro-3-picoline,2-chloro-3-methyl-5-aminopyridine,5-amino-2-chloro-3-methylpyridine,3-pyridinamine, 6-chloro-5-methyl,6-chloro-5-methyl-3-pyridylamine,6-chloro-5-methyl-pyridin-3-ylamine,6-chloro-5-methylpyridine-3-amine,pubchem1152,5-methyl-6-chloropyridine-3-amine,6-chloro-5-methyl-3-pyridinamine
12295607
6-chloro-5-methylpyridin-3-amine
CC1=CC(=CN=C1Cl)N
38186-82-2
C6H7ClN2
142.59
VSBISZPNLZFTPG-UHFFFAOYSA-N
5-amino-2-chloro-3-picoline,2-chloro-3-methyl-5-aminopyridine,5-amino-2-chloro-3-methylpyridine,3-pyridinamine, 6-chloro-5-methyl,6-chloro-5-methyl-3-pyridylamine,6-chloro-5-methyl-pyridin-3-ylamine,6-chloro-5-methylpyridine-3-amine,pubchem1152,5-methyl-6-chloropyridine-3-amine,6-chloro-5-methyl-3-pyridinamine
12295607
6-chloro-5-methylpyridin-3-amine
CC1=CC(=CN=C1Cl)N
Melting Point 90°C to 92°C
Quantity 5g
Assay Percent Range 96% min. (GC)
Formula Weight 142.59
Percent Purity 97%
Chemical Name or Material 5-Amino-2-chloro-3-picoline

GHS Signal Word: Warning