Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
5-Amino-2-chlorobenzoic acid, 97%, Acros Organics™
Click to view available options
Quantity:
25g
100g
Specifications
Quantity | 25g |
Melting Point | 184.0°C to 188.0°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 96% min. (HPLC) |
Formula Weight | 171.58 |
Percent Purity | 97% |
Chemical Name or Material | 5-Amino-2-chlorobenzoic acid |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
GHS Signal Word: Warning
EINECSNumber : 201-916-5