Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Amino-2-chlorophenol, 98%

Click to view available options
Quantity:
5g
25g
100g
6358-06-1
C6H6ClNO
143.57
MFCD01707858
JSCNCRWPXOTDDZ-UHFFFAOYSA-N
2-chloro-5-aminophenol,4-chloro-3-hydroxyaniline,phenol, 5-amino-2-chloro,5-amino-2-chloro-phenol,3-amino-6-chlorophenol,pubchem21902,3-hydroxy-4-chloroaniline,4-chloro-3-hydroxy aniline,acmc-1b64d,ksc494c4r
80676
5-amino-2-chlorophenol
C1=CC(=C(C=C1N)O)Cl
6358-06-1
C6H6ClNO
143.57
MFCD01707858
JSCNCRWPXOTDDZ-UHFFFAOYSA-N
2-chloro-5-aminophenol,4-chloro-3-hydroxyaniline,phenol, 5-amino-2-chloro,5-amino-2-chloro-phenol,3-amino-6-chlorophenol,pubchem21902,3-hydroxy-4-chloroaniline,4-chloro-3-hydroxy aniline,acmc-1b64d,ksc494c4r
80676
5-amino-2-chlorophenol
C1=CC(=C(C=C1N)O)Cl
Color Gray
Quantity 100g
Sensitivity Light Sensitive
Melting Point 160°C
UN Number UN2811
Beilstein 2081092
Formula Weight 143.57
Physical Form Powder
Percent Purity 98%
Chemical Name or Material 5-Amino-2-chlorophenol

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

RTECSNumber : SJ5720000

storageNote1 : 2° to 8°C