Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Amino-2-chlorophenol 98.0+%, TCI America™

Click to view available options
Quantity:
5g
25g
6358-06-1
C6H6ClNO
143.57
MFCD01707858
JSCNCRWPXOTDDZ-UHFFFAOYSA-N
2-chloro-5-aminophenol,4-chloro-3-hydroxyaniline,phenol, 5-amino-2-chloro,5-amino-2-chloro-phenol,3-amino-6-chlorophenol,pubchem21902,3-hydroxy-4-chloroaniline,4-chloro-3-hydroxy aniline,acmc-1b64d,ksc494c4r
80676
5-amino-2-chlorophenol
C1=CC(=C(C=C1N)O)Cl
6358-06-1
C6H6ClNO
143.57
MFCD01707858
JSCNCRWPXOTDDZ-UHFFFAOYSA-N
2-chloro-5-aminophenol,4-chloro-3-hydroxyaniline,phenol, 5-amino-2-chloro,5-amino-2-chloro-phenol,3-amino-6-chlorophenol,pubchem21902,3-hydroxy-4-chloroaniline,4-chloro-3-hydroxy aniline,acmc-1b64d,ksc494c4r
80676
5-amino-2-chlorophenol
C1=CC(=C(C=C1N)O)Cl
Quantity 25g
Color Undesignated
Melting Point 160°C
Formula Weight 143.57
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (GC,T)
Chemical Name or Material 5-Amino-2-chlorophenol

RTECSNumber : SJ5720000

TSCA : No