Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Amino-2-chloropyridine, 98%

Click to view available options
Quantity:
1g
5g
5350-93-6
C5H5ClN2
128.559
MFCD00006243
QAJYCQZQLVENRZ-UHFFFAOYSA-N
5-amino-2-chloropyridine,2-chloro-5-aminopyridine,3-amino-6-chloropyridine,3-pyridinamine, 6-chloro,6-chloro-pyridin-3-ylamine,6-chloro-3-pyridylamine,2-chloro-5-amino pyridine,6-chloro-3-pyridinamine,6-chloropyridin-3-ylamine,6-chlorpyridin-3-amin
79305
6-chloropyridin-3-amine
C1=CC(=NC=C1N)Cl
5350-93-6
C5H5ClN2
128.559
MFCD00006243
QAJYCQZQLVENRZ-UHFFFAOYSA-N
5-amino-2-chloropyridine,2-chloro-5-aminopyridine,3-amino-6-chloropyridine,3-pyridinamine, 6-chloro,6-chloro-pyridin-3-ylamine,6-chloro-3-pyridylamine,2-chloro-5-amino pyridine,6-chloro-3-pyridinamine,6-chloropyridin-3-ylamine,6-chlorpyridin-3-amin
79305
6-chloropyridin-3-amine
C1=CC(=NC=C1N)Cl
Melting Point 81°C to 84°C
Odor Unpleasant
Quantity 5g
Beilstein 108891
Formula Weight 128.56
Percent Purity 98%
Chemical Name or Material 5-Amino-2-chloropyridine

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning