Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Amino-2-chloropyrimidine, 95%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
56621-90-0
C4H4ClN3
129.55
DZBKIOJXVOECRA-UHFFFAOYSA-N
5-amino-2-chloropyrimidine,2-chloro-pyrimidin-5-ylamine,2-chloro-5-aminopyrimidine,5-pyrimidinamine, 2-chloro,2-chloropyrimidine-5-ylamine,pubchem6963,acmc-1axfh,2-chloropyrimidin-5-ylamine,5-pyrimidinamine,2-chloro,5-amino-2-chloro pyrimidine
12215993
2-chloropyrimidin-5-amine
C1=C(C=NC(=N1)Cl)N
56621-90-0
C4H4ClN3
129.55
DZBKIOJXVOECRA-UHFFFAOYSA-N
5-amino-2-chloropyrimidine,2-chloro-pyrimidin-5-ylamine,2-chloro-5-aminopyrimidine,5-pyrimidinamine, 2-chloro,2-chloropyrimidine-5-ylamine,pubchem6963,acmc-1axfh,2-chloropyrimidin-5-ylamine,5-pyrimidinamine,2-chloro,5-amino-2-chloro pyrimidine
12215993
2-chloropyrimidin-5-amine
C1=C(C=NC(=N1)Cl)N
Melting Point 160°C
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 1g
Color Yellow to Brown
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 94% min. (GC)
Formula Weight 129.55
Physical Form Crystalline Powder or Solid
Percent Purity 95%
Chemical Name or Material 5-Amino-2-chloropyrimidine
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Co

GHS Signal Word: Warning