Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Amino-2-cyanopyridine, 96%

Manufacturer:  Alfa Aesar™ H2896022

Quantity 100g

 View more versions of this product

Catalog No. AAH2896022


 

Due to product restrictions, please to purchase or view availability for this product.


Specifications

Description

Applications
Silver(I)-assisted electrophilic aromatic iodination of commercially available 5-aminopicolinonitrile 2 gave the mono-iodinated intermediate 3 in 79% yield. In mild and efficient DBU-catalyzed amidation of cyanoacetates activation of the α-carboxyl group allowed condensation with 5-amino-2-cyanopyridine.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Description & Specifications

Chemical Identifiers

55338-73-3
119.13
UN3439
5-Amino-2-cyanopyridine
C6H5N3
MFCD03093956
5-Aminopicolinonitrile; 5-Aminopyridine-2-carbonitrile

Specifications

55338-73-3
100g
119.13
UN3439
5-Amino-2-cyanopyridine
148° to 152°C
C6H5N3
MFCD03093956
5-Aminopicolinonitrile; 5-Aminopyridine-2-carbonitrile
96%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H301-H311-H319
Toxic if swallowed.
Toxic in contact with skin.
Causes serious eye irritation.

GHS P Statement
P280f-P305+P351+P338-P309-P310
Wear protective clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit