Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Amino-2-fluoro-3-methylpyridine, 95%

Click to view available options
Quantity:
250mg
1g
5g
186593-48-6
C6H7FN2
126.134
MFCD03095088
KADQKKYTJHXBSL-UHFFFAOYSA-N
5-amino-2-fluoro-3-picoline,2-fluoro-3-methyl-5-aminopyridine,5-amino-2-fluoro-3-methylpyridine,2-fluoro-5-amino-3-picoline,6-fluoro-5-methyl-3-pyridinamine,3-pyridinamine, 6-fluoro-5-methyl,6-fluoro-5-methyl-3-pyridylamine,5-amino-3-methyl-2-fluoropyridine,3-pyridinamine, 6-fluoro-5-methyl-9ci,pubchem1258
21952691
6-fluoro-5-methylpyridin-3-amine
CC1=CC(=CN=C1F)N
186593-48-6
C6H7FN2
126.134
MFCD03095088
KADQKKYTJHXBSL-UHFFFAOYSA-N
5-amino-2-fluoro-3-picoline,2-fluoro-3-methyl-5-aminopyridine,5-amino-2-fluoro-3-methylpyridine,2-fluoro-5-amino-3-picoline,6-fluoro-5-methyl-3-pyridinamine,3-pyridinamine, 6-fluoro-5-methyl,6-fluoro-5-methyl-3-pyridylamine,5-amino-3-methyl-2-fluoropyridine,3-pyridinamine, 6-fluoro-5-methyl-9ci,pubchem1258
21952691
6-fluoro-5-methylpyridin-3-amine
CC1=CC(=CN=C1F)N
Quantity 5g
Beilstein 13574091
Formula Weight 126.13
Physical Form Solid
Percent Purity 95%
Chemical Name or Material 5-Amino-2-fluoro-3-methylpyridine