Alfa Aesar™ 5-Amino-2-fluorophenol, 97%

Manufacturer: Alfa Aesar™ H33821MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH33821MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 100367-48-4
    Formula Weight 127.12
    MDL Number MFCD13185335
    Synonym 4-Fluoro-3-hydroxyaniline
    Chemical Name or Material 5-Amino-2-fluorophenol

    Specifications

    Melting Point 156° to 158°C
    Formula Weight 127.12
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Synonym 4-Fluoro-3-hydroxyaniline
    Physical Form Solid
    MDL Number MFCD13185335
    Color White to yellow
    CAS 100367-48-4
    Quantity 250mg
    Health Hazard 1 Danger
    Chemical Name or Material 5-Amino-2-fluorophenol
    Assay 97%

    Description

    Applications
    5-Amino-2-fluorophenol is used as synthetic building block.

    Notes
    Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.

    GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit