Alfa Aesar™ 5-Amino-2-fluoropyridine, 97+%

Manufacturer: Alfa Aesar™ H3087203

Quantity 1g

 View more versions of this product

Catalog No. AAH3087203

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1827-27-6
    Molecular Formula C5H5FN2
    Formula Weight 112.11
    MDL Number MFCD01632180
    UN Number UN2811
    Synonym 6-Fluoro-3-pyridinamine
    Chemical Name or Material 5-Amino-2-fluoropyridine

    Specifications

    Formula Weight 112.11
    UN Number UN2811
    Quantity 1g
    Synonym 6-Fluoro-3-pyridinamine
    Molecular Formula C5H5FN2
    Melting Point 88° to 91°C
    MDL Number MFCD01632180
    CAS 1827-27-6
    Odor Characteristic
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H312-H315-H319-H335
    Toxic if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P280h-P305+P351+P338-P309-P310
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
    Chemical Name or Material 5-Amino-2-fluoropyridine
    Assay ≥97%

    GHS H Statement
    H301-H312-H315-H319-H335
    Toxic if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P280h-P305+P351+P338-P309-P310
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit