Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Amino-2-methoxybenzoic Acid 97.0+%, TCI America™

Click to view available options
Quantity:
1g
5g
3403-47-2
C8H9NO3
167.164
MFCD09260887
LWWPSEIFAKNPKQ-UHFFFAOYSA-N
13549244
5-amino-2-methoxybenzoic acid
COC1=C(C=C(C=C1)N)C(=O)O
3403-47-2
C8H9NO3
167.164
MFCD09260887
LWWPSEIFAKNPKQ-UHFFFAOYSA-N
13549244
5-amino-2-methoxybenzoic acid
COC1=C(C=C(C=C1)N)C(=O)O
Color Red-Yellow
Melting Point 165°C
Quantity 5g
Formula Weight 167.16
Physical Form Crystal-Powder at 20°C
Percent Purity ≥97.0% (T)
Chemical Name or Material 5-Amino-2-methoxybenzoic Acid