Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Amino-2-methoxyphenol, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
10g
1687-53-2
C7H9NO2
139.154
MFCD00010222
BLQFHJKRTDIZLX-UHFFFAOYSA-N
3-hydroxy-4-methoxyaniline,5-aminoguaiacol,2-methoxy-5-aminophenol,phenol, 5-amino-2-methoxy,5-amino-2-methoxy-phenol,pubchem3112,acmc-209dyt,3-hydroxy4-methoxyaniline,5-amino-2-methoxy phenol,3-hydroxy-4-methoxylaniline
74314
5-amino-2-methoxyphenol
COC1=C(C=C(C=C1)N)O
1687-53-2
C7H9NO2
139.154
MFCD00010222
BLQFHJKRTDIZLX-UHFFFAOYSA-N
3-hydroxy-4-methoxyaniline,5-aminoguaiacol,2-methoxy-5-aminophenol,phenol, 5-amino-2-methoxy,5-amino-2-methoxy-phenol,pubchem3112,acmc-209dyt,3-hydroxy4-methoxyaniline,5-amino-2-methoxy phenol,3-hydroxy-4-methoxylaniline
74314
5-amino-2-methoxyphenol
COC1=C(C=C(C=C1)N)O
Melting Point 129°C to 133°C
Quantity 1g
Color Brown to Brown-Gray
Infrared Spectrum Authentic
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula C6H3NH(OH)OCH3
Formula Weight 139.15
Physical Form Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material 5-Amino-2-methoxyphenol, 98%
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Harmful if inhaled.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 216-873-8