Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Amino-2-methylbenzonitrile, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
50670-64-9
C8H8N2
132.17
YDZVQWCVKXYGIU-UHFFFAOYSA-N
5-amino-2-methylbenzenecarbonitrile,2-methyl-5-aminobenzonitrile,4-amino-2-cyanotoluene,3-cyano-4-methylaniline,5-amino-2-methyl-benzonitrile,benzonitrile, 5-amino-2-methyl,5-amino-o-tolunitrile,acmc-20a01k,aminomethylbenzenecarbonitrile,3-amino-6-methylbenzonitrile
2735365
5-amino-2-methylbenzonitrile
CC1=C(C=C(C=C1)N)C#N
50670-64-9
C8H8N2
132.17
YDZVQWCVKXYGIU-UHFFFAOYSA-N
5-amino-2-methylbenzenecarbonitrile,2-methyl-5-aminobenzonitrile,4-amino-2-cyanotoluene,3-cyano-4-methylaniline,5-amino-2-methyl-benzonitrile,benzonitrile, 5-amino-2-methyl,5-amino-o-tolunitrile,acmc-20a01k,aminomethylbenzenecarbonitrile,3-amino-6-methylbenzonitrile
2735365
5-amino-2-methylbenzonitrile
CC1=C(C=C(C=C1)N)C#N
Melting Point 90.9°C to 91.7°C
Quantity 5g
Infrared Spectrum Authentic
Color Yellow to Brown
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Formula Weight 132.17
Physical Form Powder
Percent Purity 97%
Chemical Name or Material 5-Amino-2-methylbenzonitrile
Show More Show Less

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
Wear protective gloves/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pr

GHS Signal Word: Warning