Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Amino-2-methylindole, 95%

Click to view available options
Quantity:
5g
Packaging:
Glass Bottle
7570-49-2
C9H10N2
146.193
MFCD02093426
JQULCCZIXYRBSE-UHFFFAOYSA-N
5-amino-2-methylindole,2-methyl-5-aminoindole,2-methylindole-5-ylamine,pubchem7273,acmc-209ozq,2-methylindole-5-amine,2-methyl-5-amino-1h-indole,bidd:gt0193,5-amino-2-methyl-1h-indole,2-methyl-1h-indol-5-ylamine
2733992
2-methyl-1H-indol-5-amine
CC1=CC2=C(N1)C=CC(=C2)N
7570-49-2
C9H10N2
146.193
MFCD02093426
JQULCCZIXYRBSE-UHFFFAOYSA-N
5-amino-2-methylindole,2-methyl-5-aminoindole,2-methylindole-5-ylamine,pubchem7273,acmc-209ozq,2-methylindole-5-amine,2-methyl-5-amino-1h-indole,bidd:gt0193,5-amino-2-methyl-1h-indole,2-methyl-1h-indol-5-ylamine
2733992
2-methyl-1H-indol-5-amine
CC1=CC2=C(N1)C=CC(=C2)N
Color Brown
Packaging Glass Bottle
Quantity 5g
Infrared Spectrum Authentic
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Formula Weight 146.19
Physical Form Crystalline Powder or Chunks
Percent Purity ≥94%
Chemical Name or Material 2-Methyl-1H-indol-5-amine
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.