Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Amino-2-methylindole, 97%

Click to view available options
Quantity:
1g
5g
25g
7570-49-2
C9H10N2
146.193
MFCD02093426
JQULCCZIXYRBSE-UHFFFAOYSA-N
5-amino-2-methylindole,2-methyl-5-aminoindole,2-methylindole-5-ylamine,pubchem7273,acmc-209ozq,2-methylindole-5-amine,2-methyl-5-amino-1h-indole,bidd:gt0193,5-amino-2-methyl-1h-indole,2-methyl-1h-indol-5-ylamine
2733992
2-methyl-1H-indol-5-amine
CC1=CC2=C(N1)C=CC(=C2)N
7570-49-2
C9H10N2
146.193
MFCD02093426
JQULCCZIXYRBSE-UHFFFAOYSA-N
5-amino-2-methylindole,2-methyl-5-aminoindole,2-methylindole-5-ylamine,pubchem7273,acmc-209ozq,2-methylindole-5-amine,2-methyl-5-amino-1h-indole,bidd:gt0193,5-amino-2-methyl-1h-indole,2-methyl-1h-indol-5-ylamine
2733992
2-methyl-1H-indol-5-amine
CC1=CC2=C(N1)C=CC(=C2)N
Quantity 25g
Formula Weight 145.19
Percent Purity 97%
Chemical Name or Material 5-Amino-2-methylindole

GHS H Statement
H301
Toxic if swallowed.

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

Recommended Storage : 2° to 8°C