Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Amino-2-methylphenol, 97%

Click to view available options
Quantity:
100g
500g
2835-95-2
C7H9NO
123.155
MFCD00043922
DBFYESDCPWWCHN-UHFFFAOYSA-N
5-amino-o-cresol,4-amino-2-hydroxytoluene,3-hydroxy-4-methylaniline,phenol, 5-amino-2-methyl,3-amino-6-methylphenol,6-methyl-3-aminophenol,unii-ubb8xeb10b,5-amino-2-methyl-phenol,5-amino-2-methyl phenol,ccris 4582
17818
5-amino-2-methylphenol
CC1=C(C=C(C=C1)N)O
2835-95-2
C7H9NO
123.155
MFCD00043922
DBFYESDCPWWCHN-UHFFFAOYSA-N
5-amino-o-cresol,4-amino-2-hydroxytoluene,3-hydroxy-4-methylaniline,phenol, 5-amino-2-methyl,3-amino-6-methylphenol,6-methyl-3-aminophenol,unii-ubb8xeb10b,5-amino-2-methyl-phenol,5-amino-2-methyl phenol,ccris 4582
17818
5-amino-2-methylphenol
CC1=C(C=C(C=C1)N)O
Melting Point 160°C to 162°C
Quantity 500g
Color Brown
Beilstein 2802317
Formula Weight 123.16
Physical Form Powder
Percent Purity 97%
Chemical Name or Material 5-Amino-2-methylphenol

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : SJ6090000

TSCA : TSCA