Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Amino-2-methylphenylboronic acid pinacol ester, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
882670-69-1
C13H20BNO2
233.12
HPNBWHDRLCVZFV-UHFFFAOYSA-N
4-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline,5-amino-2-methylphenylboronic acid, pinacol ester,5-amino-2-methylphenylboronic acid pinacol ester,4,4,5,5-tetramethyl-2-5-amino-2-methylphenyl-1,3,2-dioxaborolane,4-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzenamine,benzenamine, 4-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl,hpnbwhdrlcvzfv-uhfffaoysa-n,5-amino-2-methyl-phenylboronic acid pinacol ester,5-amino-2-methylphenyl boronic acid pinacol ester
16086089
882670-69-1
C13H20BNO2
233.12
HPNBWHDRLCVZFV-UHFFFAOYSA-N
4-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline,5-amino-2-methylphenylboronic acid, pinacol ester,5-amino-2-methylphenylboronic acid pinacol ester,4,4,5,5-tetramethyl-2-5-amino-2-methylphenyl-1,3,2-dioxaborolane,4-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzenamine,benzenamine, 4-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl,hpnbwhdrlcvzfv-uhfffaoysa-n,5-amino-2-methyl-phenylboronic acid pinacol ester,5-amino-2-methylphenyl boronic acid pinacol ester
16086089
Melting Point 83°C to 84°C
Quantity 5g
Assay Percent Range 97%
Formula Weight 233.12
Percent Purity 97%
Chemical Name or Material 5-Amino-2-methylphenylboronic acid pinacol ester

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning