Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Amino-2-(trifluoromethyl)benzimidazole, 96%, Acros Organics

Click to view available options
Quantity:
1g
5g
3671-66-7
C8H6F3N3
201.15
CKEKFQLHCAZGSP-UHFFFAOYSA-N
5-amino-2-trifluoromethyl benzimidazole,1h-benzimidazol-5-amine, 2-trifluoromethyl,2-trifluoromethyl-1h-benzo d imidazol-5-amine,benzimidazole, 5-amino-2-trifluoromethyl,2-trifluoromethyl-1h-benzimidazol-5-amine,2-trifluoromethyl-1h-1,3-benzodiazol-5-amine,5-amino-2-trifluoromethylbenzimidazole,2-trifluoromethyl benzimidazole-5-ylamine,2-trifluoromethyl-1h-benzimidazol-6-amine,2-trifluoromethyl-3h-benzoimidazol-5-ylamine
19345
2-(trifluoromethyl)-3H-benzimidazol-5-amine
C1=CC2=C(C=C1N)NC(=N2)C(F)(F)F
3671-66-7
C8H6F3N3
201.15
CKEKFQLHCAZGSP-UHFFFAOYSA-N
5-amino-2-trifluoromethyl benzimidazole,1h-benzimidazol-5-amine, 2-trifluoromethyl,2-trifluoromethyl-1h-benzo d imidazol-5-amine,benzimidazole, 5-amino-2-trifluoromethyl,2-trifluoromethyl-1h-benzimidazol-5-amine,2-trifluoromethyl-1h-1,3-benzodiazol-5-amine,5-amino-2-trifluoromethylbenzimidazole,2-trifluoromethyl benzimidazole-5-ylamine,2-trifluoromethyl-1h-benzimidazol-6-amine,2-trifluoromethyl-3h-benzoimidazol-5-ylamine
19345
2-(trifluoromethyl)-3H-benzimidazol-5-amine
C1=CC2=C(C=C1N)NC(=N2)C(F)(F)F
Packaging Glass Bottle
Quantity 1g
Melting Point 189.0°C to 191.0°C
Assay Percent Range 95% min. (HPLC)
Formula Weight 201.15
Percent Purity 96%
Chemical Name or Material 5-Amino-2-(trifluoromethyl)benzimidazole

GHS H Statement
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for

GHS Signal Word: Danger