Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Amino-3-methyl-1-p-tolylpyrazole 97.0+%, TCI America™

Click to view available options
Quantity:
10g
25g
62535-60-8
C11H13N3
187.246
MFCD00020725
WQUNBEPKWJLYJD-UHFFFAOYSA-N
112855
5-methyl-2-(4-methylphenyl)pyrazol-3-amine
CC1=CC=C(C=C1)N2C(=CC(=N2)C)N
62535-60-8
C11H13N3
187.246
MFCD00020725
WQUNBEPKWJLYJD-UHFFFAOYSA-N
112855
5-methyl-2-(4-methylphenyl)pyrazol-3-amine
CC1=CC=C(C=C1)N2C(=CC(=N2)C)N
Color Undesignated
Melting Point 118°C
Quantity 25g
Formula Weight 187.25
Physical Form Crystal-Powder at 20°C
Percent Purity ≥97.0% (GC,T)
Chemical Name or Material 5-Amino-3-methyl-1-p-tolylpyrazole