Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Amino-4,6-dichloropyrimidine, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
5413-85-4
C4H3Cl2N3
163.99
MFCD00006108
NIGDWBHWHVHOAD-UHFFFAOYSA-N
5-amino-4,6-dichloropyrimidine,5-pyrimidinamine, 4,6-dichloro,4,6-dichloro-5-pyrimidinamine,4,6-dichloro-5-aminopyrimidine,4,6-dichloropyrimidin-5-ylamine,5-amine-4,6-dichloropyrimidine,5-amino-4,6-dichloropyfimidine,4,6-dichloro-pyrimidin-5-ylamine,pyrimidine, 5-amino-4,6-dichloro,4,6-dichloropyrimidine-5-ylamine
79434
4,6-dichloropyrimidin-5-amine
C1=NC(=C(C(=N1)Cl)N)Cl
5413-85-4
C4H3Cl2N3
163.99
MFCD00006108
NIGDWBHWHVHOAD-UHFFFAOYSA-N
5-amino-4,6-dichloropyrimidine,5-pyrimidinamine, 4,6-dichloro,4,6-dichloro-5-pyrimidinamine,4,6-dichloro-5-aminopyrimidine,4,6-dichloropyrimidin-5-ylamine,5-amine-4,6-dichloropyrimidine,5-amino-4,6-dichloropyfimidine,4,6-dichloro-pyrimidin-5-ylamine,pyrimidine, 5-amino-4,6-dichloro,4,6-dichloropyrimidine-5-ylamine
79434
4,6-dichloropyrimidin-5-amine
C1=NC(=C(C(=N1)Cl)N)Cl
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 25g
Melting Point 143°C to 148°C
Color Brown-Yellow
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (HPLC)
Solubility Information Solubility in water: slightly soluble
Formula Weight 163.99
Physical Form Crystals or Powder
Percent Purity 97%
Chemical Name or Material 5-Amino-4, 6-dichloropyrimidine
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

EINECSNumber : 226-503-7