Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Amino-4,6-dichloropyrimidine, 98+%

Click to view available options
Quantity:
1g
5g
25g
5413-85-4
C4H3Cl2N3
163.989
MFCD00006108
NIGDWBHWHVHOAD-UHFFFAOYSA-N
5-amino-4,6-dichloropyrimidine,5-pyrimidinamine, 4,6-dichloro,4,6-dichloro-5-pyrimidinamine,4,6-dichloro-5-aminopyrimidine,4,6-dichloropyrimidin-5-ylamine,5-amine-4,6-dichloropyrimidine,5-amino-4,6-dichloropyfimidine,4,6-dichloro-pyrimidin-5-ylamine,pyrimidine, 5-amino-4,6-dichloro,4,6-dichloropyrimidine-5-ylamine
79434
4,6-dichloropyrimidin-5-amine
C1=NC(=C(C(=N1)Cl)N)Cl
5413-85-4
C4H3Cl2N3
163.989
MFCD00006108
NIGDWBHWHVHOAD-UHFFFAOYSA-N
5-amino-4,6-dichloropyrimidine,5-pyrimidinamine, 4,6-dichloro,4,6-dichloro-5-pyrimidinamine,4,6-dichloro-5-aminopyrimidine,4,6-dichloropyrimidin-5-ylamine,5-amine-4,6-dichloropyrimidine,5-amino-4,6-dichloropyfimidine,4,6-dichloro-pyrimidin-5-ylamine,pyrimidine, 5-amino-4,6-dichloro,4,6-dichloropyrimidine-5-ylamine
79434
4,6-dichloropyrimidin-5-amine
C1=NC(=C(C(=N1)Cl)N)Cl
Melting Point 143°C to 146°C
Quantity 1g
Odor Irritating
Beilstein 126885
Formula Weight 164
Percent Purity ≥98%
Chemical Name or Material 5-Amino-4,6-dichloropyrimidine

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA