Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Amino-4-chloropyrimidine, 96%

Click to view available options
Quantity:
250mg
1g
5g
54660-78-5
C4H4ClN3
129.547
MFCD06412561
LHGMCUVJFRBVBH-UHFFFAOYSA-N
5-amino-4-chloropyrimidine,4-chloro-5-aminopyrimidine,4-chloranylpyrimidin-5-amine,5-pyrimidinamine, 4-chloro,4-chloro-5-amino-pyrimidine,acmc-209lid,4-chloro-5-pyrimidinamine,5-amino-6-chloropyrimidine,4-chloropyrimidine-5-amine,4-chloropyrimidin-5-ylamine
13404541
4-chloropyrimidin-5-amine
C1=C(C(=NC=N1)Cl)N
54660-78-5
C4H4ClN3
129.547
MFCD06412561
LHGMCUVJFRBVBH-UHFFFAOYSA-N
5-amino-4-chloropyrimidine,4-chloro-5-aminopyrimidine,4-chloranylpyrimidin-5-amine,5-pyrimidinamine, 4-chloro,4-chloro-5-amino-pyrimidine,acmc-209lid,4-chloro-5-pyrimidinamine,5-amino-6-chloropyrimidine,4-chloropyrimidine-5-amine,4-chloropyrimidin-5-ylamine
13404541
4-chloropyrimidin-5-amine
C1=C(C(=NC=N1)Cl)N
Quantity 5g
Color Brown to White
Melting Point 110°C to 116°C
Sensitivity Air & Light Sensitive
Formula Weight 129.55
Physical Form Solid
Percent Purity 96%
Chemical Name or Material 5-Amino-4-chloropyrimidine

storageNote1 : 2° to 8°C