Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Amino-6-chloro-2-picoline, 98+%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
39745-40-9
C6H7ClN2
142.59
MFCD03095213
SUMSBUDMYMYTLL-UHFFFAOYSA-N
3-amino-2-chloro-6-picoline,3-amino-2-chloro-6-methylpyridine,5-amino-6-chloro-2-picoline,2-chloro-6-methyl-pyridin-3-ylamine,2-chloro-3-amino-6-methylpyridine,3-pyridinamine, 2-chloro-6-methyl,2-chloro-6-methyl-3-pyridylamine,pubchem1245,acmc-20dld6,2-chloro-3-amino-6-picoline
2734415
2-chloro-6-methylpyridin-3-amine
CC1=NC(=C(C=C1)N)Cl
39745-40-9
C6H7ClN2
142.59
MFCD03095213
SUMSBUDMYMYTLL-UHFFFAOYSA-N
3-amino-2-chloro-6-picoline,3-amino-2-chloro-6-methylpyridine,5-amino-6-chloro-2-picoline,2-chloro-6-methyl-pyridin-3-ylamine,2-chloro-3-amino-6-methylpyridine,3-pyridinamine, 2-chloro-6-methyl,2-chloro-6-methyl-3-pyridylamine,pubchem1245,acmc-20dld6,2-chloro-3-amino-6-picoline
2734415
2-chloro-6-methylpyridin-3-amine
CC1=NC(=C(C=C1)N)Cl
Melting Point 74°C to 80°C
Color Cream to Tan
Packaging Glass bottle
Quantity 1g
Infrared Spectrum Authentic
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98% min. (HPLC)
Formula Weight 142.59
Physical Form Crystalline Powder
Percent Purity 98+%
Chemical Name or Material 5-Amino-6-chloro-2-picoline
Show More Show Less

GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Harmful if inhaled.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning