Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Amino-N,N′-bis(2,3-dihydroxypropyl)isophthalamide Hydrochloride 98.0+%, TCI America™

Click to view available options
Quantity:
5g
25g
203515-86-0
C14H22ClN3O6
363.795
GNBBCFFLJFKAHB-UHFFFAOYSA-N
44629992
5-amino-1-N,3-N-bis(2,3-dihydroxypropyl)benzene-1,3-dicarboxamide;hydrochloride
C1=C(C=C(C=C1C(=O)NCC(CO)O)N)C(=O)NCC(CO)O.Cl
203515-86-0
C14H22ClN3O6
363.795
GNBBCFFLJFKAHB-UHFFFAOYSA-N
44629992
5-amino-1-N,3-N-bis(2,3-dihydroxypropyl)benzene-1,3-dicarboxamide;hydrochloride
C1=C(C=C(C=C1C(=O)NCC(CO)O)N)C(=O)NCC(CO)O.Cl
Quantity 25g
Color White-Yellow
Formula Weight 363.80
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (HPLC,N)
Chemical Name or Material 5-Amino-N,N′-bis(2,3-dihydroxypropyl)isophthalamide Hydrochloride