Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Aminobenzene-1,3-diol hydrochloride, Technical Grade, Maybridge™

Click to view available options
Quantity:
250mg
1g
5g
6318-56-5
C6H8ClNO2
161.585
VNZZCDQPCQIUGG-UHFFFAOYSA-N
5-aminobenzene-1,3-diol hydrochloride, 1,3-benzenediol, 5-amino-, hydrochloride, 5-aminoresorcinol, hydrochloride, pubchem19962, 5-aminobenzene-1,3-diol hcl, 5-aminobenzene-1,3-diol hydrochloridyl
459248
5-aminobenzene-1,3-diol;hydrochloride
C1=C(C=C(C=C1O)O)N.Cl
6318-56-5
C6H8ClNO2
161.585
VNZZCDQPCQIUGG-UHFFFAOYSA-N
5-aminobenzene-1,3-diol hydrochloride, 1,3-benzenediol, 5-amino-, hydrochloride, 5-aminoresorcinol, hydrochloride, pubchem19962, 5-aminobenzene-1,3-diol hcl, 5-aminobenzene-1,3-diol hydrochloridyl
459248
5-aminobenzene-1,3-diol;hydrochloride
C1=C(C=C(C=C1O)O)N.Cl
Packaging Amber Glass Bottle
Quantity 1g
Linear Formula NH2C6H3(OH)2.HCl
Formula Weight 161.59
Percent Purity Tech.
Grade Technical
Chemical Name or Material 5-Aminobenzene-1, 3-diol hydrochloride