Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Aminoisophthalic acid, 98%, ACROS Organics™

missing translation for 'orderingAttributeHoverText'
Quantity:
25g
100g
Packaging:
Plastic bottle
99-31-0
C8H7NO4
181.15
MFCD00002522
KBZFDRWPMZESDI-UHFFFAOYSA-N
5-aminoisophthalic acid, 1,3-benzenedicarboxylic acid, 5-amino, 5-amino-1,3-isophthalic acid, isophthalic acid, 5-amino, 5-amino-1,3-benzenedicarboxylic acid, 3,5-dicarboxyaniline, 5-aminoisophtalic acid, 3,5-dicarboxy-aniline, pubchem20539, anhydrous
66833
5-aminobenzene-1,3-dicarboxylic acid
C1=C(C=C(C=C1C(=O)O)N)C(=O)O
99-31-0
C8H7NO4
181.15
MFCD00002522
KBZFDRWPMZESDI-UHFFFAOYSA-N
5-aminoisophthalic acid, 1,3-benzenedicarboxylic acid, 5-amino, 5-amino-1,3-isophthalic acid, isophthalic acid, 5-amino, 5-amino-1,3-benzenedicarboxylic acid, 3,5-dicarboxyaniline, 5-aminoisophtalic acid, 3,5-dicarboxy-aniline, pubchem20539, anhydrous
66833
5-aminobenzene-1,3-dicarboxylic acid
C1=C(C=C(C=C1C(=O)O)N)C(=O)O
Melting Point >300°C
Infrared Spectrum Authentic
Quantity 100g
Packaging Plastic bottle
Water 6% max.
Color White to Cream
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 97.5% min. (HPLC)
Beilstein 14, I, 636
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol
Formula Weight 181.15
Physical Form Granular Powder
Percent Purity 98%
Chemical Name or Material 5-Aminoisophthalic acid, 98+%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 202-748-5

TSCA : TSCA