Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Aminonicotinic acid, 95%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
24242-19-1
C6H6N2O2
138.13
MFCD00129116
BYIORJAACCWFPU-UHFFFAOYSA-N
5-aminonicotinic acid,5-amino-nicotinic acid,5-aminonicotinicacid,3-pyridinecarboxylic acid, 5-amino,5-amino nicotinic acid,5-amino-3-carboxypyridine,5-amino-3-pyridinecarboxylic acid,zlchem 307,pubchem15914,acmc-209ga2
354316
CHEBI:68578
5-aminopyridine-3-carboxylic acid
C1=C(C=NC=C1N)C(=O)O
24242-19-1
C6H6N2O2
138.13
MFCD00129116
BYIORJAACCWFPU-UHFFFAOYSA-N
5-aminonicotinic acid,5-amino-nicotinic acid,5-aminonicotinicacid,3-pyridinecarboxylic acid, 5-amino,5-amino nicotinic acid,5-amino-3-carboxypyridine,5-amino-3-pyridinecarboxylic acid,zlchem 307,pubchem15914,acmc-209ga2
354316
CHEBI:68578
5-aminopyridine-3-carboxylic acid
C1=C(C=NC=C1N)C(=O)O
Color Brown
Packaging Glass bottle
Quantity 5g
Infrared Spectrum Authentic
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 94% min. (HPLC)
Formula Weight 138.13
Physical Form Crystalline Powder
Percent Purity 95%
Chemical Name or Material 5-Aminonicotinic acid
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning