Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Aminopyrazine-2-carboxylic acid, 95%

Click to view available options
Quantity:
250mg
1g
5g
40155-43-9
C5H5N3O2
139.114
MFCD07371664
FMEFOOOBIHQRMK-UHFFFAOYSA-N
5-amino-2-pyrazinecarboxylic acid,2-amino-5-pyrazinecarboxylic acid,pyrazinecarboxylic acid, 5-amino,5-amino-pyrazine-2-carboxylic acid,5-aminopyrazine-2-carboxylicacid,5-amino-pyrazine-2-carboxylic acid,5-amino-pyrazine-2-carboxylicacid,pubchem7866,5-amino-pyrazinecarboxylic acid,2-amino-5-pirazinecarboxylic acid
21897334
5-aminopyrazine-2-carboxylic acid
C1=C(N=CC(=N1)N)C(=O)O
40155-43-9
C5H5N3O2
139.114
MFCD07371664
FMEFOOOBIHQRMK-UHFFFAOYSA-N
5-amino-2-pyrazinecarboxylic acid,2-amino-5-pyrazinecarboxylic acid,pyrazinecarboxylic acid, 5-amino,5-amino-pyrazine-2-carboxylic acid,5-aminopyrazine-2-carboxylicacid,5-amino-pyrazine-2-carboxylic acid,5-amino-pyrazine-2-carboxylicacid,pubchem7866,5-amino-pyrazinecarboxylic acid,2-amino-5-pirazinecarboxylic acid
21897334
5-aminopyrazine-2-carboxylic acid
C1=C(N=CC(=N1)N)C(=O)O
Quantity 1g
Beilstein 120673
Formula Weight 139.11
Physical Form Solid
Percent Purity 95%
Chemical Name or Material 5-Aminopyrazine-2-carboxylic acid

storageNote1 : -30° to -10°C