Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-(Benzyloxycarbonylamino)valeric acid, 97%

Click to view available options
Quantity:
250mg
1g
5g
23135-50-4
C13H17NO4
251.282
MFCD00188458
QYYPKLYDFCYGPG-UHFFFAOYSA-N
5-benzyloxy carbonyl amino pentanoic acid,5-benzyloxy carbonyl amino pentanoic acid,5-benzyloxycarbonylamino-pentanoic acid,5-benzyloxycarbonylamino pentanoic acid,z-5-ava-oh,5-carbobenzoxyamino valeric acid,5-carbobenzoxyamino pentanoic acid,5-phenylmethoxycarbonylamino pentanoic acid,5-benzyloxycarbonylamino-pentanoicacid,pubchem20738
3690024
5-(phenylmethoxycarbonylamino)pentanoic acid
C1=CC=C(C=C1)COC(=O)NCCCCC(=O)O
23135-50-4
C13H17NO4
251.282
MFCD00188458
QYYPKLYDFCYGPG-UHFFFAOYSA-N
5-benzyloxy carbonyl amino pentanoic acid,5-benzyloxy carbonyl amino pentanoic acid,5-benzyloxycarbonylamino-pentanoic acid,5-benzyloxycarbonylamino pentanoic acid,z-5-ava-oh,5-carbobenzoxyamino valeric acid,5-carbobenzoxyamino pentanoic acid,5-phenylmethoxycarbonylamino pentanoic acid,5-benzyloxycarbonylamino-pentanoicacid,pubchem20738
3690024
5-(phenylmethoxycarbonylamino)pentanoic acid
C1=CC=C(C=C1)COC(=O)NCCCCC(=O)O
Melting Point 99°C to 105°C
Color White
Quantity 250mg
Linear Formula C6H4CH2OCONH2(CH2)4CO2H
Beilstein 2334766
Formula Weight 251.28
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 5-(Benzyloxycarbonylamino)valeric acid