Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Benzyloxyindole-3-carboxaldehyde, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
6953-22-6
C16H13NO2
251.28
MFCD00014562
DJGNUBADRQIDNQ-UHFFFAOYSA-N
5-benzyloxyindole-3-carboxaldehyde,5-benzyloxyindole-3-carbaldehyde,5-benzyloxy-1h-indole-3-carbaldehyde,5-phenylmethoxy-1h-indole-3-carbaldehyde,5-benzyloxy-3-formylindole,5-benzyloxy-indole-3-carboxaldehyde,5-benzyloxy-3-formyl-1h-indole,5-benzyloxy-3-indolecarboxaldehyde,5-benzyloxy indole-3-carboxyaldehyde
81398
5-phenylmethoxy-1H-indole-3-carbaldehyde
C1=CC=C(C=C1)COC2=CC3=C(C=C2)NC=C3C=O
6953-22-6
C16H13NO2
251.28
MFCD00014562
DJGNUBADRQIDNQ-UHFFFAOYSA-N
5-benzyloxyindole-3-carboxaldehyde,5-benzyloxyindole-3-carbaldehyde,5-benzyloxy-1h-indole-3-carbaldehyde,5-phenylmethoxy-1h-indole-3-carbaldehyde,5-benzyloxy-3-formylindole,5-benzyloxy-indole-3-carboxaldehyde,5-benzyloxy-3-formyl-1h-indole,5-benzyloxy-3-indolecarboxaldehyde,5-benzyloxy indole-3-carboxyaldehyde
81398
5-phenylmethoxy-1H-indole-3-carbaldehyde
C1=CC=C(C=C1)COC2=CC3=C(C=C2)NC=C3C=O
Melting Point 241°C to 242°C
Quantity 1g
Packaging Glass bottle
Color Yellow to Brown
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (HPLC)
Formula Weight 251.28
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material 5-Benzyloxyindole-3-carboxaldehyde
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

EINECSNumber : 230-134-7