Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Benzyloxyindole, 94%, may contain up to ca 7% toluene

Click to view available options
Quantity:
1g
5g
25g
1215-59-4
C15H13NO
223.275
MFCD00005676
JCQLPDZCNSVBMS-UHFFFAOYSA-N
5-benzyloxyindole,5-benzyloxy-1h-indole,1h-indole, 5-phenylmethoxy,benzyloxy-5 indole,indole, 5-benzyloxy,5-benzyloxy indole,unii-yci4z02e1c
14624
5-phenylmethoxy-1H-indole
C1=CC=C(C=C1)COC2=CC3=C(C=C2)NC=C3
1215-59-4
C15H13NO
223.275
MFCD00005676
JCQLPDZCNSVBMS-UHFFFAOYSA-N
5-benzyloxyindole,5-benzyloxy-1h-indole,1h-indole, 5-phenylmethoxy,benzyloxy-5 indole,indole, 5-benzyloxy,5-benzyloxy indole,unii-yci4z02e1c
14624
5-phenylmethoxy-1H-indole
C1=CC=C(C=C1)COC2=CC3=C(C=C2)NC=C3
Melting Point 96°C to 101°C
Quantity 25g
Beilstein 173532
Formula Weight 223.28
Percent Purity 94%
Chemical Name or Material 5-Benzyloxyindole
Assay may contain up to ≈7% toluene

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : NL4850000