Alfa Aesar™ 5-Benzyloxytryptamine hydrochloride, 98%

Manufacturer: Alfa Aesar™ H5108903

Quantity 1g

 View more versions of this product

Catalog No. AAH5108903

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 52055-23-9
    Molecular Formula C17H18N2O·HCl
    Formula Weight 302.8
    MDL Number MFCD00012685
    Synonym 3-(2-Aminoethyl)-5-benzyloxyindole hydrochloride
    Chemical Name or Material 5-Benzyloxytryptamine hydrochloride

    Specifications

    Formula Weight 302.8
    Molecular Formula C17H18N2O·HCl
    MDL Number MFCD00012685
    Synonym 3-(2-Aminoethyl)-5-benzyloxyindole hydrochloride
    Melting Point 259° to 263°C
    Quantity 1g
    CAS 52055-23-9
    Chemical Name or Material 5-Benzyloxytryptamine hydrochloride
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit