Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-(beta-Styryl)-2H-1,2,3,4-tetrazole, 99%

Manufacturer:  Alfa Aesar™ H5110706

Quantity 5g

 View more versions of this product

Catalog No. AAH5110706


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

220429-71-0
172.19
6502443
5-(beta-Styryl)-2H-1,2,3,4-tetrazole
C9H8N4
MFCD00276928
5-(2-Phenylethenyl)-2H-tetraazole

Specifications

220429-71-0
172.19
6502443
5g
99%
C9H8N4
MFCD00276928
5-(2-Phenylethenyl)-2H-tetraazole
5-(beta-Styryl)-2H-1,2,3,4-tetrazole
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit