Alfa Aesar™ 5-(beta-Styryl)-2H-1,2,3,4-tetrazole, 99%

Manufacturer: Alfa Aesar™ H5110703

Quantity 1g

 View more versions of this product

Catalog No. AAH5110703

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 220429-71-0
    Molecular Formula C9H8N4
    Formula Weight 172.19
    MDL Number MFCD00276928
    Beilstein 6502443
    Synonym 5-(2-Phenylethenyl)-2H-tetraazole
    Chemical Name or Material 5-(beta-Styryl)-2H-1,2,3,4-tetrazole

    Specifications

    Formula Weight 172.19
    Molecular Formula C9H8N4
    Synonym 5-(2-Phenylethenyl)-2H-tetraazole
    MDL Number MFCD00276928
    CAS 220429-71-0
    Beilstein 6502443
    Quantity 1g
    Chemical Name or Material 5-(beta-Styryl)-2H-1,2,3,4-tetrazole
    Assay 99%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit