Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-(Boc-amino)valeric acid, 95%

Click to view available options
Quantity:
1g
5g
25g
27219-07-4
C10H19NO4
217.265
MFCD00076903
GFMRZAMDGJIWRB-UHFFFAOYSA-N
boc-5-ava-oh,5-tert-butoxycarbonylamino valeric acid,boc-5-aminovaleric acid,5-boc-amino valeric acid,5-tert-butoxycarbonyl amino pentanoic acid,5-boc-amino-pentanoic acid,5-tert-butoxycarbonyl amino pentanoic acid,n-boc-5-aminopentanoic acid,pentanoic acid, 5-1,1-dimethylethoxy carbonyl amino,n-boc-5-aminovaleric acid
545848
5-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid
CC(C)(C)OC(=O)NCCCCC(=O)O
27219-07-4
C10H19NO4
217.265
MFCD00076903
GFMRZAMDGJIWRB-UHFFFAOYSA-N
boc-5-ava-oh,5-tert-butoxycarbonylamino valeric acid,boc-5-aminovaleric acid,5-boc-amino valeric acid,5-tert-butoxycarbonyl amino pentanoic acid,5-boc-amino-pentanoic acid,5-tert-butoxycarbonyl amino pentanoic acid,n-boc-5-aminopentanoic acid,pentanoic acid, 5-1,1-dimethylethoxy carbonyl amino,n-boc-5-aminovaleric acid
545848
5-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid
CC(C)(C)OC(=O)NCCCCC(=O)O
Quantity 5g
Boiling Point 160°C to 168°C (0.8mmHg)
Melting Point 48°C to 52°C
Color White
Linear Formula (CH3)3COC(=O)NH(CH2)4CO2H
Beilstein 2048862
Formula Weight 217.27
Physical Form Powder
Percent Purity 95%
Chemical Name or Material 5-(Boc-amino)valeric acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning