Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Bromo-1,2,3-trimethoxybenzene, 97%

Click to view available options
Quantity:
5g
25g
100g
2675-79-8
C9H11BrO3
247.088
MFCD00017169
XAOOZMATJDXDQJ-UHFFFAOYSA-N
1-bromo-3,4,5-trimethoxybenzene,3,4,5-trimethoxybromobenzene,bromotrimethoxybenzene,benzene, 5-bromo-1,2,3-trimethoxy,5-bromo-1,2,3-trimethoxy-benzene,pubchem10980,3,4,5-trimethoxybromoben,ksc490o1d,3,4,5-trimethoxyphenylbromide,3,4,5-triemethoxyphenylbromide
75885
5-bromo-1,2,3-trimethoxybenzene
COC1=CC(=CC(=C1OC)OC)Br
2675-79-8
C9H11BrO3
247.088
MFCD00017169
XAOOZMATJDXDQJ-UHFFFAOYSA-N
1-bromo-3,4,5-trimethoxybenzene,3,4,5-trimethoxybromobenzene,bromotrimethoxybenzene,benzene, 5-bromo-1,2,3-trimethoxy,5-bromo-1,2,3-trimethoxy-benzene,pubchem10980,3,4,5-trimethoxybromoben,ksc490o1d,3,4,5-trimethoxyphenylbromide,3,4,5-triemethoxyphenylbromide
75885
5-bromo-1,2,3-trimethoxybenzene
COC1=CC(=CC(=C1OC)OC)Br
Applications
5-Bromo-1,2,3-trimethoxybenzene is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Quantity 25g
Melting Point 77°C to 80°C
Formula Weight 247.1
Percent Purity 97%
Chemical Name or Material 5-Bromo-1,2,3-trimethoxybenzene

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning