Alfa Aesar™ 5-Bromo-1,2,4-trimethylbenzene, 99%

Manufacturer: Alfa Aesar™ B2327114

Quantity 25g

 View more versions of this product

Catalog No. AAB2327114

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 5469-19-2
    Molecular Formula C9H11Br
    Formula Weight 199.09
    MDL Number MFCD00000072
    Synonym 2,4,5-Trimethylbromobenzene
    Chemical Name or Material 5-Bromo-1,2,4-trimethylbenzene

    Specifications

    Formula Weight 199.09
    Molecular Formula C9H11Br
    MDL Number MFCD00000072
    CAS 5469-19-2
    Melting Point 71° to 73°C
    Synonym 2,4,5-Trimethylbromobenzene
    Boiling Point 233° to 235°C
    Quantity 25g
    Chemical Name or Material 5-Bromo-1,2,4-trimethylbenzene
    Assay 99%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit