Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-1-methyl-1H-indole-3-carboxylic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
400071-95-6
C10H8BrNO2
254.08
MFCD14706162
WQZLFFJMRKTCMU-UHFFFAOYSA-N
5-bromo-1-methyl-1h-indole-3-carboxylic acid,5-bromo-1-methyl-1h-indole-3-carboxylicacid,1h-indole-3-carboxylic acid, 5-bromo-1-methyl,ksc230c9b,5-bromo-1-methyl-1h-indole-3-carboxylic acid;
11032413
5-bromo-1-methylindole-3-carboxylic acid
CN1C=C(C2=C1C=CC(=C2)Br)C(=O)O
400071-95-6
C10H8BrNO2
254.08
MFCD14706162
WQZLFFJMRKTCMU-UHFFFAOYSA-N
5-bromo-1-methyl-1h-indole-3-carboxylic acid,5-bromo-1-methyl-1h-indole-3-carboxylicacid,1h-indole-3-carboxylic acid, 5-bromo-1-methyl,ksc230c9b,5-bromo-1-methyl-1h-indole-3-carboxylic acid;
11032413
5-bromo-1-methylindole-3-carboxylic acid
CN1C=C(C2=C1C=CC(=C2)Br)C(=O)O
Packaging Glass bottle
Quantity 5g
Infrared Spectrum Authentic
Melting Point 255.4°C to 255.7°C
Color White
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (HPLC)
Formula Weight 254.08
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material 5-Bromo-1-methyl-1H-indole-3-carboxylic acid
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

GHS Signal Word: Warning