Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-1-methylimidazole, 97%, Acros Organics

Click to view available options
Quantity:
5g
25g
1003-21-0
C4H5BrN2
161
HATLLUIOEIXWGD-UHFFFAOYSA-N
5-bromo-1-methyl-1h-imidazole,5-bromo-n-methylimidazole,pubchem8951,1h-imidazole, 5-bromo-1-methyl,5-bromo-1-methyl-imidazole,imidazole,5-bromo-1-methyl,5-bromo-n-methyl-imidazole,5-bromo-1-methyl imidazole,acmc-1c6k3,1-methyl-5-bromo-1h-imidazole
1515258
5-bromo-1-methylimidazole
CN1C=NC=C1Br
1003-21-0
C4H5BrN2
161
HATLLUIOEIXWGD-UHFFFAOYSA-N
5-bromo-1-methyl-1h-imidazole,5-bromo-n-methylimidazole,pubchem8951,1h-imidazole, 5-bromo-1-methyl,5-bromo-1-methyl-imidazole,imidazole,5-bromo-1-methyl,5-bromo-n-methyl-imidazole,5-bromo-1-methyl imidazole,acmc-1c6k3,1-methyl-5-bromo-1h-imidazole
1515258
5-bromo-1-methylimidazole
CN1C=NC=C1Br
Boiling Point 120°C (15.0mmHg)
Melting Point 40°C to 41°C
Flash Point 82°C
Packaging Glass bottle
Quantity 25g
Assay Percent Range 96% min. (GC)
Formula Weight 161
Percent Purity 97%
Chemical Name or Material 5-Bromo-1-methylimidazole

GHS Signal Word: Danger