Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Bromo-1-pentene, 96%

Manufacturer:  Alfa Aesar™ A1121903

Quantity 1g

 View more versions of this product

Catalog No. AAA1121903


  Check Availability
Add to cart

Specifications

Description

Applications
5-Bromo-1-pentene is widely utilized in the stereoselective synthesis of 7alpha-(3-carboxypropyl) estradiol. It plays an important role as starting material in the synthesis of a variety of compounds including DL-histrionicotoxin and benzophenone containing fatty acids.

Solubility
Immiscible with water.

Notes
Keep away from sources of ignition.
Description & Specifications

Chemical Identifiers

1119-51-3
Br(CH2)3CH=CH2
MFCD0000-364
506077
BrC5H9
149.03
UN1993
5-Bromo-1-pentene

Specifications

1119-51-3
1g
BrC5H9
149.03
UN1993
5-Bromo-1-pentene
126° to 127°C
30°C (86°F)
Br(CH2)3CH=CH2
MFCD0000-364
506077
96%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

TSCA : TSCA

Recommended Storage : 2°C to 8°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit