Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-1H-pyrazolo[3,4-b]pyridine, 95%, Alfa Aesar™

Click to view available options
Quantity:
250mg
1g
875781-17-2
C6H4BrN3
198.023
MFCD05663982
BASYLPMLKGQZOG-UHFFFAOYSA-N
5-bromo-1h-pyrazolo 3,4-b pyridine,5-bromo-2h-pyrazolo 3,4-b pyridine,5-bromo-1h-pyrazo 3,4-b pyridine,1h-pyrazolo 3,4-b pyridine, 5-bromo,5-bromo-7-azaindazole,pubchem17877,acmc-209qoi,7-aza-5-bromo-1h-indole,ksc496o5j,5-bromopyrazolo 3,4-b pyridine
40427454
5-bromo-1H-pyrazolo[3,4-b]pyridine
C1=C2C=NNC2=NC=C1Br
875781-17-2
C6H4BrN3
198.023
MFCD05663982
BASYLPMLKGQZOG-UHFFFAOYSA-N
5-bromo-1h-pyrazolo 3,4-b pyridine,5-bromo-2h-pyrazolo 3,4-b pyridine,5-bromo-1h-pyrazo 3,4-b pyridine,1h-pyrazolo 3,4-b pyridine, 5-bromo,5-bromo-7-azaindazole,pubchem17877,acmc-209qoi,7-aza-5-bromo-1h-indole,ksc496o5j,5-bromopyrazolo 3,4-b pyridine
40427454
5-bromo-1H-pyrazolo[3,4-b]pyridine
C1=C2C=NNC2=NC=C1Br
Quantity 1g
Beilstein 11434713
Formula Weight 198.02
Physical Form Solid
Percent Purity 95%
Chemical Name or Material 5-Bromo-1H-pyrazolo[3,4-b]pyridine