Learn More
5-Bromo-2,2-difluoro-1,3-benzodioxole, 97%, Thermo Scientific Chemicals
Description
5-Bromo-2,2-difluoro-1,3- benzodioxole is used as a precursor in organic synthesis. It is used to prepare 5-bromo-2,2-difluoro-1,3-benzodioxole-4-carboxylic acid. It serves as an intermediate in active pharmaceutical ingredients, agrochemicals and in dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
5-Bromo-2,2-difluoro-1,3- benzodioxole is used as a precursor in organic synthesis. It is used to prepare 5-bromo-2,2-difluoro-1,3-benzodioxole-4-carboxylic acid. It serves as an intermediate in active pharmaceutical ingredients, agrochemicals and in dyestuff field.
Solubility
Miscible with hexane.
Notes
Light sensitive. Incompatible with strong oxidizing agents and strong acids.
Specifications
Density | 1.74 |
Boiling Point | 78°C to 79°C (20 mmHg) |
Refractive Index | 1.4967 |
Quantity | 5 g |
Beilstein | 1425209 |
Solubility Information | Miscible with hexane. |
Formula Weight | 237 |
Percent Purity | 97% |
Chemical Name or Material | 5-Bromo-2,2-difluoro-1,3-benzodioxole |
Safety and Handling
Hazard Category | H315-H319 |
Hazard Statement | GHS H Statement H315-H319 Causes skin irritation. Causes serious eye irritation. |
Precautionary Statement | P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362 |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.