Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Bromo-2,3-difluorophenol, 97%

Click to view available options
Quantity:
1g
5g
25g
186590-26-1
C6H3BrF2O
208.99
MFCD01631350
QAHCQGXGAYRHHW-UHFFFAOYSA-N
2,3-difluoro-5-bromophenol,phenol, 5-bromo-2,3-difluoro,5-bromo-2,3-difluoro-phenol,3-bromo-5,6-difluorophenol,pubchem2843,acmc-209eoh,ksc494o6r,5-bromo-2,3-difluoro phenol,phenol,5-bromo-2,3-difluoro,2.3-difluoro-5-bromophenol
2773307
5-bromo-2,3-difluorophenol
C1=C(C=C(C(=C1F)F)O)Br
186590-26-1
C6H3BrF2O
208.99
MFCD01631350
QAHCQGXGAYRHHW-UHFFFAOYSA-N
2,3-difluoro-5-bromophenol,phenol, 5-bromo-2,3-difluoro,5-bromo-2,3-difluoro-phenol,3-bromo-5,6-difluorophenol,pubchem2843,acmc-209eoh,ksc494o6r,5-bromo-2,3-difluoro phenol,phenol,5-bromo-2,3-difluoro,2.3-difluoro-5-bromophenol
2773307
5-bromo-2,3-difluorophenol
C1=C(C=C(C(=C1F)F)O)Br
Quantity 1g
Beilstein 8831044
Formula Weight 208.99
Percent Purity 97%
Chemical Name or Material 5-Bromo-2,3-difluorophenol

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger