Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Bromo-2,3-difluoropyridine, 97%

Click to view available options
Quantity:
1g
5g
25g
89402-44-8
C5H2BrF2N
193.979
MFCD06659525
QVIQXJRQVOPYGI-UHFFFAOYSA-N
pyridine, 5-bromo-2,3-difluoro,2,3-difluoro-5-bromopyridine,pubchem1292,5-bromo-2,3-difluoro-pyridine,pyridine,5-bromo-2,3-difluoro,5-bromanyl-2,3-bis fluoranyl pyridine
14549654
5-bromo-2,3-difluoropyridine
C1=C(C(=NC=C1Br)F)F
89402-44-8
C5H2BrF2N
193.979
MFCD06659525
QVIQXJRQVOPYGI-UHFFFAOYSA-N
pyridine, 5-bromo-2,3-difluoro,2,3-difluoro-5-bromopyridine,pubchem1292,5-bromo-2,3-difluoro-pyridine,pyridine,5-bromo-2,3-difluoro,5-bromanyl-2,3-bis fluoranyl pyridine
14549654
5-bromo-2,3-difluoropyridine
C1=C(C(=NC=C1Br)F)F
Applications
It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Quantity 1g
Color Colorless to Yellow
Beilstein 13412561
Formula Weight 193.98
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 5-Bromo-2,3-difluoropyridine

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning