Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Bromo-2,4-dichloropyrimidine, 98%

Click to view available options
Quantity:
1g
5g
36082-50-5
C4HBrCl2N2
227.87
MFCD00127818
SIKXIUWKPGWBBF-UHFFFAOYSA-N
2,4-dichloro-5-bromopyrimidine,2,4-dichloro-5-bromo pyrimidine,5-bromo-2,4-dichloro-pyrimidine,pyrimidine, 5-bromo-2,4-dichloro,5-bromo-2,6-dichloropyrimidine,5-bromo-2,4-dichloro-1,3-diazine,5-brom-2,4-dichlorpyrimidin,zlchem 380,pubchem3993,acmc-1ad68
289973
5-bromo-2,4-dichloropyrimidine
C1=C(C(=NC(=N1)Cl)Cl)Br
36082-50-5
C4HBrCl2N2
227.87
MFCD00127818
SIKXIUWKPGWBBF-UHFFFAOYSA-N
2,4-dichloro-5-bromopyrimidine,2,4-dichloro-5-bromo pyrimidine,5-bromo-2,4-dichloro-pyrimidine,pyrimidine, 5-bromo-2,4-dichloro,5-bromo-2,6-dichloropyrimidine,5-bromo-2,4-dichloro-1,3-diazine,5-brom-2,4-dichlorpyrimidin,zlchem 380,pubchem3993,acmc-1ad68
289973
5-bromo-2,4-dichloropyrimidine
C1=C(C(=NC(=N1)Cl)Cl)Br
Boiling Point 128°C (15mmHg)
Melting Point 29°C to 30°C
Flash Point >110°C (230°F)
Quantity 1g
UN Number UN2923
Beilstein 124441
Formula Weight 227.88
Percent Purity 98%
Chemical Name or Material 5-Bromo-2,4-dichloropyrimidine

GHS H Statement
H301-H314-H318-H317
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.

GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.