Alfa Aesar™ 5-Bromo-2,4-dichloropyrimidine, 98%

Manufacturer: Alfa Aesar™ B2536506

Quantity 5g

 View more versions of this product

Catalog No. AAB2536506

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 36082-50-5
    Molecular Formula C4HBrCl2N2
    Formula Weight 227.88
    MDL Number MFCD00127818
    UN Number UN2923
    Beilstein 124441
    Chemical Name or Material 5-Bromo-2,4-dichloropyrimidine

    Specifications

    Formula Weight 227.88
    CAS 36082-50-5
    Boiling Point 128°C (15mmHg)
    Quantity 5g
    Flash Point >110°C (230°F)
    Health Hazard 2 GHS H Statement
    H301-H314-H318-H317
    Toxic if swallowed.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    May cause an allergic skin reaction.
    Molecular Formula C4HBrCl2N2
    MDL Number MFCD00127818
    UN Number UN2923
    Melting Point 29° to 30°C
    Beilstein 124441
    Health Hazard 1 Danger
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
    Chemical Name or Material 5-Bromo-2,4-dichloropyrimidine
    Assay 98%

    GHS H Statement
    H301-H314-H318-H317
    Toxic if swallowed.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    May cause an allergic skin reaction.

    GHS P Statement
    P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit