Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-2,4-dichloropyrimidine, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
36082-50-5
C4HBrCl2N2
227.88
MFCD00127818
SIKXIUWKPGWBBF-UHFFFAOYSA-N
2,4-dichloro-5-bromopyrimidine,2,4-dichloro-5-bromo pyrimidine,5-bromo-2,4-dichloro-pyrimidine,pyrimidine, 5-bromo-2,4-dichloro,5-bromo-2,6-dichloropyrimidine,5-bromo-2,4-dichloro-1,3-diazine,5-brom-2,4-dichlorpyrimidin,zlchem 380,pubchem3993,acmc-1ad68
289973
5-bromo-2,4-dichloropyrimidine
C1=C(C(=NC(=N1)Cl)Cl)Br
36082-50-5
C4HBrCl2N2
227.88
MFCD00127818
SIKXIUWKPGWBBF-UHFFFAOYSA-N
2,4-dichloro-5-bromopyrimidine,2,4-dichloro-5-bromo pyrimidine,5-bromo-2,4-dichloro-pyrimidine,pyrimidine, 5-bromo-2,4-dichloro,5-bromo-2,6-dichloropyrimidine,5-bromo-2,4-dichloro-1,3-diazine,5-brom-2,4-dichlorpyrimidin,zlchem 380,pubchem3993,acmc-1ad68
289973
5-bromo-2,4-dichloropyrimidine
C1=C(C(=NC(=N1)Cl)Cl)Br
Specific Gravity 1.781
Quantity 5g
Infrared Spectrum Authentic
Packaging Glass bottle
Color Colorless to Yellow
Melting Point 29.0°C to 30.0°C
Boiling Point 128.0°C (15.0 mmHg)
Density 1.7810g/mL
Refractive Index 1.6020 to 1.6040 (20°C, 589nm)
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Formula Weight 227.88
Physical Form Liquid or Low Melting Solid
Percent Purity 98%
Chemical Name or Material 5-Bromo-2, 4-dichloropyrimidine
Show More Show Less

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger