Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-2,4-dihydroxybenzoic acid monohydrate, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
160348-98-1
C7H5BrO4·H2O
251.03
MFCD00149094
MRIKDYLDHHINRD-UHFFFAOYSA-N
5-bromo-2,4-dihydroxybenzoic acid monohydrate,5-bromo-2,4-dihydroxybenzoic acid hydrate,zlchem 1095,c7h5bro4.h2o,acmc-1c3u5,5-bromo-2,4-dihydroxybenzoic acid, hydrate,5-bromo-2,4-dihydroxybenzoic acid-water 1/1,benzoic acid,5-bromo-2,4-dihydroxy-,hydrate 1:1
16211702
5-bromo-2,4-dihydroxybenzoic acid;hydrate
C1=C(C(=CC(=C1Br)O)O)C(=O)O.O
160348-98-1
C7H5BrO4·H2O
251.03
MFCD00149094
MRIKDYLDHHINRD-UHFFFAOYSA-N
5-bromo-2,4-dihydroxybenzoic acid monohydrate,5-bromo-2,4-dihydroxybenzoic acid hydrate,zlchem 1095,c7h5bro4.h2o,acmc-1c3u5,5-bromo-2,4-dihydroxybenzoic acid, hydrate,5-bromo-2,4-dihydroxybenzoic acid-water 1/1,benzoic acid,5-bromo-2,4-dihydroxy-,hydrate 1:1
16211702
5-bromo-2,4-dihydroxybenzoic acid;hydrate
C1=C(C(=CC(=C1Br)O)O)C(=O)O.O
Melting Point 209°C to 210°C
Color White to Yellow
Quantity 5g
Infrared Spectrum Authentic
Water 7.8% max.
Packaging Glass bottle
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (HPLC)
Linear Formula BrC6H2(OH)2CHO
Formula Weight 251.03
Physical Form Powder
Percent Purity 97%
Chemical Name or Material 5-Bromo-2, 4-dihydroxybenzoic acid monohydrate
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning