Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-2-chloro-3-nitropyridine, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
67443-38-3
C5H2BrClN2O2
237.44
MFCD00222270
WWQQPSDIIVXFOX-UHFFFAOYSA-N
2-chloro-3-nitro-5-bromopyridine,5-bromo-2-chloro-3-nitro-pyridine,pyridine, 5-bromo-2-chloro-3-nitro,abbypharma ap-12-5159,zlchem 393,pubchem1166,acmc-209nz1,ksc352q2l,5-bromo-2-chloro-3-nitropyridin,2-chloro-5-bromo-3-nitropyridine
7019412
5-bromo-2-chloro-3-nitropyridine
C1=C(C(=NC=C1Br)Cl)[N+](=O)[O-]
67443-38-3
C5H2BrClN2O2
237.44
MFCD00222270
WWQQPSDIIVXFOX-UHFFFAOYSA-N
2-chloro-3-nitro-5-bromopyridine,5-bromo-2-chloro-3-nitro-pyridine,pyridine, 5-bromo-2-chloro-3-nitro,abbypharma ap-12-5159,zlchem 393,pubchem1166,acmc-209nz1,ksc352q2l,5-bromo-2-chloro-3-nitropyridin,2-chloro-5-bromo-3-nitropyridine
7019412
5-bromo-2-chloro-3-nitropyridine
C1=C(C(=NC=C1Br)Cl)[N+](=O)[O-]
Melting Point 64.0°C to 69.0°C
Color Yellow
Infrared Spectrum Authentic
Quantity 5g
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (HPLC)
Formula Weight 237.44
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material 5-Bromo-2-chloro-3-nitropyridine
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning