Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-2-chloro-3-picoline, 98%, Acros Organics™

Click to view available options
Quantity:
5g
25g
29241-60-9
C6H5BrClN
206.47
YTSKHJMMNFPBBZ-UHFFFAOYSA-N
2-chloro-3-methyl-5-bromopyridine,5-bromo-2-chloro-3-picoline,5-bromo-2-chloro-3-methyl pyridine,5-bromo-2-chloro-3-methyl-pyridine,pyridine, 5-bromo-2-chloro-3-methyl,2-chloro-5-bromo-3-picoline,2-chloro-3-methyl-5-bromo pyridine,pubchem1104,ksc495o9j
285434
5-bromo-2-chloro-3-methylpyridine
CC1=CC(=CN=C1Cl)Br
29241-60-9
C6H5BrClN
206.47
YTSKHJMMNFPBBZ-UHFFFAOYSA-N
2-chloro-3-methyl-5-bromopyridine,5-bromo-2-chloro-3-picoline,5-bromo-2-chloro-3-methyl pyridine,5-bromo-2-chloro-3-methyl-pyridine,pyridine, 5-bromo-2-chloro-3-methyl,2-chloro-5-bromo-3-picoline,2-chloro-3-methyl-5-bromo pyridine,pubchem1104,ksc495o9j
285434
5-bromo-2-chloro-3-methylpyridine
CC1=CC(=CN=C1Cl)Br
Quantity 25g
Melting Point 40.0°C to 46.0°C
Infrared Spectrum Authentic
Boiling Point 78.0°C (3.0 mmHg)
Assay Percent Range 97.5% min. (GC)
Beilstein 20,V,5,538
Formula Weight 206.47
Percent Purity 98%
Chemical Name or Material 5-Bromo-2-chloro-3-picoline

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning